Odlingsrör
14 ml, PP, med lock, Styckvis, 95 mm, 17 mm
Katalog # 720-0565
Leverantör: Simport Scientific
Gå till:
Product Details & Documents
PP eller PS, graderad, engångs, steril.
- White marking field
- Gradation distinguishing 0,1 (4 ml tube) and 0,25 ml (12 ml tube)
- Biological inert
- Durable, reduces the risk in handling potentially hazardous material
Passar för de flesta rutinapplikationer.
Resist temperatures from −190 to +120 °C (PP) and up to 70 °C (PS).
Max. RCF: 3000×g (PP); 1400×g (PS).
Leveransinformation: Levereras med eller utan lock av PE.
Documents
Specifikationer
- Ø:17 mm
- Förp.:Styckvis
- Kapacitet:14 ml
- Beskrivning:PP, med lock
- Höjd:95 mm
- Seq:11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



