Orders LinkContactUsLinkComponent
Test strips
Test strips
Alkalinity (free and total CaCO₃), 0 - 240 mg/L, TS, 50
Katalog # 705-0185
Leverantör:   Hach
Test strips
Katalog # 705-0185
Leverantör:  Hach
CAS-nummer:  
Test strips
Test strips
Alkalinity (free and total CaCO₃), 0 - 240 mg/L, TS, 50
CAS-nummer:  
Price
435,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 50 Items
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
Documents
Specifikationer
Specifikationer
  • Förp.:
    50
  • Beskrivning:
    Alkalinity (free and total CaCO₃)
  • Typ:
    TS
  • Mätområde:
    0 - 240 mg/L