Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution, up to 200 mg/l SO₃, 1 SET
Katalog # HACH2440000
Leverantör:   Hach
Test kits, drop count tests
Katalog # HACH2440000
Leverantör:  Hach
CAS-nummer:  
Test kits, drop count tests
Test kits, drop count tests
Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution, up to 200 mg/l SO₃, 1 SET
CAS-nummer:  
Price
1 950,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 SET
Produktdetaljer och dokument
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifikationer
Specifikationer
  • Förp.:
    1 SET
  • Beskrivning:
    Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution
  • Mätområde:
    up to 200 mg/l SO₃