Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0,0493 N titrant), 1 SET
Katalog # HACH2437600
Leverantör:   Hach
Test kits, drop count tests
Katalog # HACH2437600
Leverantör:  Hach
CAS-nummer:  
Test kits, drop count tests
Test kits, drop count tests
Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0,0493 N titrant), 1 SET
CAS-nummer:  
Price
1 550,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 SET
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Varning: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifikationer
Specifikationer
  • Förp.:
    1 SET
  • Beskrivning:
    Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0,0493 N titrant)
Specifikationer