Orders LinkContactUsLinkComponent
Large Rectangular Waste Bins
Large Rectangular Waste Bins
310 L, 900×810×590 mm
Catalog # CORAG145821
Supplier:   Gilac PROFESSIONNEL
Large Rectangular Waste Bins
Catalog # CORAG145821
Large Rectangular Waste Bins
310 L, 900×810×590 mm
Price
13 375,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
LDPE.
  • Mechanical and chemical resistance
  • Durability and versatility
  • Easy to clean
Specifications
  • Kapacitet:
    310 L
  • Seq:
    05
  • B×D×H:
    900×810×590 mm