Large Rectangular Waste Bins
220 L, 790×720×525 mm
Katalog # CORAG145621
Leverantör: Gilac PROFESSIONNEL
Gå till:
Produktdetaljer och dokument
LDPE.
- Mechanical and chemical resistance
- Durability and versatility
- Easy to clean
Specifikationer
- Kapacitet:220 L
- Seq:04
- B×D×H:790×720×525 mm