Large Rectangular Waste Bins
100 L, 595×555×475 mm
Catalog # 216-2880
Supplier: Gilac PROFESSIONNEL
Jump to:
Product Details & Documents
LDPE.
- Mechanical and chemical resistance
- Durability and versatility
- Easy to clean
Specifications
- Kapacitet:100 L
- Seq:02
- B×D×H:595×555×475 mm