Orders LinkContactUsLinkComponent
Test kits, drop count tests
Chloride, model 8-P, use silver nitrate titrant, 5 - 100 mg/l Cl-
20 - 400 mg/l Cl-, 1 KIT
Catalog # HACH144001
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH144001
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests
Chloride, model 8-P, use silver nitrate titrant, 5 - 100 mg/l Cl-
20 - 400 mg/l Cl-, 1 KIT
CAS Number:  
Price
1 625,00 kr
Log in for availability
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Förp.:
    1 KIT
  • Beskrivning:
    Chloride, model 8-P, use silver nitrate titrant
  • Mätområde:
    5 - 100 mg/l Cl-<br />20 - 400 mg/l Cl-