Natriumklorid ≥98%, TECHNICAL
Hink (plast), 5 kg
Leverantör: VWR Chemicals
Gå till:
Product Details & Documents
Synonymer: Saline, NaCl, Sea salt, Halite
- CAS-nummer:7647-14-5
- Formel:NaCl
- MDL-nummer:MFCD00003477
- Molecular Weight:58.44 g/mol
- Merck Index:14,08599
- Kokande Pt:1413 °C (1013 hPa)
- Smält Pt:801 °C
- Densitet:2,16 g/cm³ (20 °C)
- Förvaringstemperatur:Omgivande
- EINECS:231-598-3
Documents
Certificates
Sök efter LOT-nummer1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
P20_VWRC27788.297_SPEC_EN.pdf P20_VWRC27788.366_SPEC_EN.pdf P20_VWRC27788.460_SPEC_EN.pdf spec_VWRC27788.297 spec_VWRC27788.297
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifikationer
Specifikationstestresultat
- Assay:Min. 98 %
- Appearance:Colourless crystals
Specifikationer
- Förpackningstyp:Hink (plast)
- Förp.:5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



