Sample cuvettes
Sample cuvettes, with lid, matched pair, 10/25 ml marked, 25 ml, 1" / 25 mm, Optiskt glas
Katalog # HACH2612602
Leverantör: Hach
Gå till:
Product Details & Documents
Sample cuvettes for photometric measurements.
- Available with or without lid
- Available reusable (made from glass) or for single use (made from plastics)
Specifikationer
- Skiktdjup:1" / 25 mm
- Volym:25 ml
- Material:Optiskt glas
- Beskrivning:Sample cuvettes, with lid, matched pair, 10/25 ml marked
- Seq:10