Ampoule breakers
Ampoule breaker, 2 ml PourRite ampoules
Katalog # HACH2484600
Leverantör: Hach
Gå till:
Produktdetaljer och dokument
These breakers for ampoules are designed to prevent contamination. Breaker snaps ampoule tip off.
Specifikationer
- Beskrivning:Ampoule breaker
- Seq:01
- För:2 ml PourRite ampoules