Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
60 ml, 14/23 NS
Katalog # BIBB443038N
Leverantör:   SciLabware
Separating funnels, Gilson
Katalog # BIBB443038N
Leverantör:  SciLabware
Separating funnels, Gilson
Separating funnels, Gilson
60 ml, 14/23 NS
Price
1 075,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.
Specifikationer
  • Kärna:
    14/23 NS
  • Kapacitet:
    60 ml
  • Seq:
    08