ContactUsLinkComponent
GASTUBE CHLORIDE IN SOLN. 25-1000 MG/L 1 * 10 Items
Katalog # GAST221L
Leverantör:  GASTEC
GASTUBE CHLORIDE IN SOLN. 25-1000 MG/L 1 * 10 Items
Katalog # GAST221L
Leverantör:  GASTEC