Order Entry
Sweden
Orders LinkContactUsLinkComponent
Reducing pieces for NW flanges

Reducing pieces for NW flanges

Reducing piece, 28 mm, NW 25/16
Katalog # EDWAC10514447
Reducing pieces for NW flanges
Katalog # EDWAC10514447
Reducing pieces for NW flanges

Reducing piece, 28 mm, NW 25/16

Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents

Reducing pieces made of stainless steel.

Certifications: AISI 316L DIN 1.4404

Specifikationer
  • Beskrivning:
    Reducing piece, 28 mm, NW 25/16
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.