Orders LinkContactUsLinkComponent
Droppflaskor, WHEATON®
15-415, Gul
Katalog # 215-4018
Leverantör:   DWK Life Sciences
Droppflaskor, WHEATON®
Katalog # 215-4018
Leverantör:  DWK Life Sciences
Droppflaskor, WHEATON®
Droppflaskor, WHEATON®
15-415, Gul
Price
600,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifikationer
  • Gänga:
    15-415
  • Seq:
    31
  • Lock, färg:
    Gul