Droppflaskor, WHEATON®
15-415, Röd
Katalog # 215-2764
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gänga:15-415
- Seq:25
- Lock, färg:Röd