Droppflaskor, WHEATON®
8-425, Grön
Katalog # 215-4009
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gänga:8-425
- Seq:06
- Lock, färg:Grön