Orders LinkContactUsLinkComponent
Droppflaskor, WHEATON®
Droppflaskor, WHEATON®
8-425, Röd
Catalog # 215-4005
Supplier:   DWK Life Sciences
Droppflaskor, WHEATON®
Catalog # 215-4005
Supplier:  DWK Life Sciences
Droppflaskor, WHEATON®
Droppflaskor, WHEATON®
8-425, Röd
Price
565,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifications
  • Gänga:
    8-425
  • Seq:
    02
  • Lock, färg:
    Röd