Screw caps, bakelite
with Butyl/PTFE seal, 40 mm, Svart, Bakelit
Katalog # 215-2685
Leverantör: GRAVIS
Gå till:
Product Details & Documents
Documents
SPVAP08028_TDS_EN
Datablad
Specifikationer
- Gänga:40 mm
- Färg:Svart
- Beskrivning:with Butyl/PTFE seal
- Material:Bakelit
- Seq:07