BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), un-numbered, 300 ml
Katalog # WHEA227497-00G
Leverantör: DWK Life Sciences
Gå till:
Produktdetaljer och dokument
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specifikationer
- Kapacitet:300 ml
- Beskrivning:BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), un-numbered