Vacuum stopcocks
Vacuum stopcocks, two way, right-angled, 2,5 mm, 14,5 NS
Katalog # LENZ02961402
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
- Excellent chemical resistance
- Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Hål Ø.:2,5 mm
- Beskrivning:Vacuum stopcocks, two way, right-angled
- Seq:16
- Kran:14,5 NS