Order Entry
Sweden
Orders LinkContactUsLinkComponent
EJ SE

EJ SE

Sterican®, för neural terapi, 23, 80 mm, 0,60 mm, Blå
Catalog # 612-0162
Supplier:   B. Braun
EJ SE
Catalog # 612-0162
Supplier:  B. Braun
EJ SE
EJ SE

Sterican®, för neural terapi, 23, 80 mm, 0,60 mm, Blå

Price
450,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Stainless, chromium-nickel steel with transparent PP Luer lock plastic hub. These sterile hypodermic needles Sterican® have the following features: Suitable for single-use hypodermic needles for intra-muscular, subcutaneous, intravenous, intra-arterial and special injections. Thin walled needles.

  • Extremely smooth surface with light silicone coating
  • 3-facet tips, long or short bevel
  • For minimal pain upon puncture

Certifications: ISO 6009

Specifications
  • Färgkod:
    Blå
  • Ø:
    0,60 mm
  • Mätare:
    23
  • Beskrivning:
    Sterican®, för neural terapi
  • Längd:
    80 mm
  • Seq:
    23