Analogue stopwatches, split-second addition timer
Analogt stoppur, ⅒ sekund, 15 minuter, tryckknapp och sju stenar, 130 grams
Katalog # 609-1132
Leverantör: HANHART
Gå till:
Product Details & Documents
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
- Chrome plated brass casing, diamond cut
- Lever, 7 jewels
- Shock resistant, additional dust and water protection
Specifikationer
- Beskrivning:Analogt stoppur, ⅒ sekund, 15 minuter, tryckknapp och sju stenar, 130 grams