Wire baskets
540×255×60 mm
Katalog # 216-7452
Leverantör: HUPFER METALLWERKE
Gå till:
Product Details & Documents
Rostfritt stål.
- Two flip handles
- Base frame is stackable
- Excellent thermal and chemical resistance
Wire steriliser strainer basket.
Specifikationer
- B×D×H:540×255×60 mm