Dropptratt
100 ml, 29/32 NS, 29/32 NS, 2 ml
201-1126
DURAN®, borosilicate glass, clear, with PTFE key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
: DIN ISO 4800
- Kärna:29/32 NS
- NS-hylsa:29/32 NS
- Kapacitet:100 ml
- Indelning:2 ml



