Proppar, rombisk form
19/26
Katalog # 217-0802
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Flat
- Diamond-ground
Specifikationer
- NS:19/26
- Seq:08