Proppar, rombisk form
10/19
Katalog # 217-0797
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Flat
- Diamond-ground
Specifikationer
- NS:10/19
- Seq:03