Proppar, rombisk form
7/16
Katalog # 217-0796
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Flat
- Diamond-ground
Specifikationer
- NS:7/16
- Seq:01
DURAN®, borosilicate glass 3.3, clear.