Orders LinkContactUsLinkComponent
ORP electrode solutions
ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 640 mV, 60 Ampoul
Katalog # SCOI285138381
Leverantör:   SI ANALYTICS
ORP electrode solutions
Katalog # SCOI285138381
Leverantör:  SI ANALYTICS
CAS-nummer:  
ORP electrode solutions
ORP electrode solutions
ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 640 mV, 60 Ampoul
CAS-nummer:  
Price
3 050,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 60 Ampoul
Product Details & Documents
ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.
Specifikationer
Specifikationer
  • Förp.:
    60 Ampoul
  • Beskrivning:
    ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 640 mV
Specifikationer