Kran, sidoställd
Stopcock with PTFE key, byretter 25 och 50 ml kapacitet
Catalog # BRND81927
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 5,4, amber.
- Interchangeable key
- With glass or PTFE key
- Key with retention device
Stopcock housing with burette tip and key.
Specifications
- För:byretter 25 och 50 ml kapacitet
- Beskrivning:Stopcock with PTFE key
- Seq:03