Densitetsflaskor, okalibrerade
100 ml
Katalog # 614-0344
Leverantör: Brand
Gå till:
Produktdetaljer och dokument
Borosilicate 3.3 glass, Gay-Lussac type
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
- Nominal capacity printed on the bottom
Certifications: DIN ISO 3507
Specifikationer
- Volym (nominell):100 ml