Orders LinkContactUsLinkComponent
Densitetsflaskor, okalibrerade
50 ml
  614-0343
 :   Brand
Densitetsflaskor, okalibrerade
  614-0343
 :  Brand
Densitetsflaskor, okalibrerade
Densitetsflaskor, okalibrerade
50 ml
1 225,00 kr
1 * 2 Items
Borosilicate 3.3 glass, Gay-Lussac type
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • Nominal capacity printed on the bottom
 : DIN ISO 3507
  • Volym (nominell):
    50 ml