Densitetsflaskor, okalibrerade
5 ml
614-0339
: Brand
Borosilicate 3.3 glass, Gay-Lussac type
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
- Nominal capacity printed on the bottom
: DIN ISO 3507
- Volym (nominell):5 ml
Borosilicate 3.3 glass, Gay-Lussac type
: DIN ISO 3507