Accessories
Thread adapter for 5 l jerrycan from 45 mm to 17/8" thread
Catalog # EPPE4960832.006
Supplier:   EPPENDORF
Accessories
Catalog # EPPE4960832.006
Supplier:  EPPENDORF
Accessories
Accessories
Thread adapter for 5 l jerrycan from 45 mm to 17/8" thread
Price
785,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivning:
    Thread adapter for 5 l jerrycan from 45 mm to 17/8" thread