Orders LinkContactUsLinkComponent
Presskruv
Presskruv
Presskruv, röd
Katalog # 554-1148
Leverantör:   VICI JOUR RESEARCH
Presskruv
Katalog # 554-1148
Leverantör:  VICI JOUR RESEARCH
Presskruv
Presskruv
Presskruv, röd
Price
760,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 6 Items
Product Details & Documents
PEEK one-piece fingertight fittings. Work with all ¹/₁₆'' OD tubings and female 10 to 32 ports. They are rated to 350 bar and can be used with virtually any HPLC fittings, eg. Swagelok, Parker, Rheodyne, Valco.
  • 100% inert and biocompatible
  • Same colour coding as capillary tubing
Specifikationer
  • Beskrivning:
    Presskruv, röd