Kiselgel ≥99.0%
Glass bottle for solids, 5 kg
Purity: 99.5% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Fused Amorphous Powder, S.A. 5m2/g Density: 2.2 pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- :7631-86-9
- :SiO₂
- :MFCD00011232
- :60.08 g/mol
- :14,8493
- :2230 °C
- :1710 °C
- :2,2
- :Omgivande
- :231-545-4
- Förpackningstyp:Glass bottle for solids
- Förp.:5 kg