Orders LinkContactUsLinkComponent
Kiselgel ≥99.0%
Glass bottle for solids, 1 kg
  89709.A1
Kiselgel ≥99.0%
  89709.A1
 :  7631-86-9
Kiselgel ≥99.0%
Kiselgel ≥99.0%
Glass bottle for solids, 1 kg
 :  7631-86-9
5 450,00 kr
1 * 1 kg
Purity: 99.5% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Fused Amorphous Powder, S.A. 5m2/g Density: 2.2 pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
 :  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  •  :7631-86-9
  •  :SiO₂
  •  :MFCD00011232
  •  :60.08 g/mol
  •  :14,8493
  •  :2230 °C
  •  :1710 °C
  •  :2,2
  •  :Omgivande
  •  :231-545-4
  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    1 kg