Kiselgel ≥99.995%
Glass bottle for solids, 25 g
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
Purity: 99.995% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -40 Mesh Powder Density: 2.2 Appearance: White Odour: Odourless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
Synonymer: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- CAS-nummer:7631-86-9
- Formel:SiO₂
- MDL-nummer:MFCD00011232
- Molecular Weight:60.08 g/mol
- Merck Index:14,8493
- Kokande Pt:2230 °C
- Smält Pt:1710 °C
- Densitet:2,2
- Förvaringstemperatur:Omgivande
- EINECS:231-545-4
Specifikationer
Specifikationer
- Förpackningstyp:Glass bottle for solids
- Förp.:25 g



