Order Entry
Sweden
Orders LinkContactUsLinkComponent
Kiselgel ≥99.5%

Kiselgel ≥99.5%

Glass bottle for solids, 1 kg
Katalog # 88316.A1
Hälsa
Leverantör:   Thermo Fisher Scientific
Kiselgel ≥99.5%
Katalog # 88316.A1
CAS-nummer:  14808-60-7
Kiselgel ≥99.5%
Kiselgel ≥99.5%

Glass bottle for solids, 1 kg

CAS-nummer:  14808-60-7
Price
5 400,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 kg
Product Details & Documents

Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

Purity: 99.5% CAS No.: 14808-60-7 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Powder Density: 2.2-2.6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Synonymer:  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  • CAS-nummer:14808-60-7
  • Formel:SiO₂
  • MDL-nummer:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Merck Index:14,8493
  • Kokande Pt:2230 °C
  • Smält Pt:1710 °C
  • Densitet:2,2…2.6
  • Förvaringstemperatur:Omgivande
  • EINECS:238-878-4
Specifikationer

Specifikationer

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.