Orders LinkContactUsLinkComponent
Cellskrapa
Cellskrapa
Cellskrapa med 18 mm vridbart blad, 400 mm
Katalog # 391-3011
Leverantör:   Greiner Bio-One
Cellskrapa
Katalog # 391-3011
Leverantör:  Greiner Bio-One
Cellskrapa
Cellskrapa
Cellskrapa med 18 mm vridbart blad, 400 mm
Price
5 500,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents
Designed to facilitate the gentle mechanical removal of adherent cells from growth surfaces. Each scraper has a 18 mm pivoted (pivot angle 60°) scraper blade, which allows access to poorly accessible surfaces.
  • Optimised blade design for maximum cell harvest
  • Uniform contact with the growth surface, minimal mechanical strain and efficient cell harvest, even from poorly accessible surfaces
  • Minimises any accumulation of the cell suspension to the blade structure
Förpackning: Individuellt förpackade.
Documents
Specifikationer
  • Beskrivning:
    Cellskrapa med 18 mm vridbart blad
  • Total längd:
    400 mm