Orders LinkContactUsLinkComponent
Filter discs
Filter discs
50,8 mm, 3,18 mm
Katalog # 42299.KS
Leverantör:   Thermo Fisher Scientific
Filter discs
Katalog # 42299.KS
Filter discs
50,8 mm, 3,18 mm
Price
1 400,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
These quartz discs are manufactured by fusing naturally occurring crystalline silica.
  • Ground and polished
Specifikationer
  • Ø:
    50,8 mm
  • Seq:
    02
  • Tjocklek:
    3,18 mm