Fused quartz microscope slides
Microscope slides, Fused quartz, 76×26 mm
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These fused quartz slides are made of fusing naturally occurring crystalline silica.
- High chemical purity, high corrosion resistance, high melting point
- Low coefficient of thermal expansion with excellent electrical insulation
- Optical transmission from ultra-violet to infrared
Documents
Specifications
- Beskrivning:Microscope slides
- L×B:76×26 mm
- Material:Fused quartz
- Seq:03