Fused quartz microscope slides
Microscope slides, Fused quartz, 26×51 mm
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
These fused quartz slides are made of fusing naturally occurring crystalline silica.
- High chemical purity, high corrosion resistance, high melting point
- Low coefficient of thermal expansion with excellent electrical insulation
- Optical transmission from ultra-violet to infrared
Documents
Specifikationer
- Beskrivning:Microscope slides
- L×B:26×51 mm
- Material:Fused quartz
- Seq:02