Order Entry
Sweden
Orders LinkContactUsLinkComponent
Kiselgel ≥99.99%

Kiselgel ≥99.99%

Glass bottle for solids, 100 g
Katalog # 36346.22
Leverantör:   Thermo Fisher Scientific
Kiselgel ≥99.99%
Katalog # 36346.22
CAS-nummer:  7631-86-9
Kiselgel ≥99.99%
Kiselgel ≥99.99%

Glass bottle for solids, 100 g

CAS-nummer:  7631-86-9
Price
4 825,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 g
Produktdetaljer och dokument

Purity: 99.99% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 3-12mm Fused Lump Density: 2.2-2.6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Synonymer:  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  • CAS-nummer:7631-86-9
  • Formel:SiO₂
  • MDL-nummer:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Merck Index:14,8493
  • Kokande Pt:2230 °C
  • Smält Pt:1710 °C
  • Densitet:2,2…2.6
  • Förvaringstemperatur:Omgivande
  • EINECS:231-545-4
Specifikationer

Specifikationer

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.