Order Entry
Sweden
Orders LinkContactUsLinkComponent
Vanadium(IV) oxide ≥99%

Vanadium(IV) oxide ≥99%

Glass bottle for solids, 10 g
Katalog # 22957.09
Irriterande
Leverantör:   Thermo Fisher Scientific
Vanadium(IV) oxide ≥99%
Katalog # 22957.09
CAS-nummer:  12036-21-4
Vanadium(IV) oxide ≥99%
Vanadium(IV) oxide ≥99%

Glass bottle for solids, 10 g

CAS-nummer:  12036-21-4
Price
2 150,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 10 g
Produktdetaljer och dokument

Purity: 99% CAS No.: 12036-21-4 Molecular Formula: O2V, may contain V6O13 Formula Weight: 82.94 Physical Form: -100 Mesh Powder Density: 4.339 Appearance: Black Melting Point: 1967° UN No.: 3285 MDL No.: MFCD00049702

Synonymer:  Vanadium dioxide
  • CAS-nummer:12036-21-4
  • Formel:O₂V
  • MDL-nummer:MFCD00049702
  • Molecular Weight:82.94 g/mol
  • Kokande Pt:2700 °C (1013 hPa)
  • Smält Pt:1967 °C
  • Densitet:4,339
  • Förvaringstemperatur:Omgivande
  • EINECS:234-841-1
  • FN:3285
  • ADR:6.1,III
  •  :Density: 4,33 g/cm³ (20 °C)
Specifikationer

Specifikationer

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    10 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.