Order Entry
Sweden
Orders LinkContactUsLinkComponent
Ammoniumformat ≥98%

Ammoniumformat ≥98%

Plastic bottle for solids, 250 g
Catalog # 14517.30
Irritant
Ammoniumformat ≥98%
Catalog # 14517.30
CAS Number:  540-69-2
Ammoniumformat ≥98%
Ammoniumformat ≥98%

Plastic bottle for solids, 250 g

CAS Number:  540-69-2
Price
2 950,00 kr
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 540-69-2 Molecular Formula: CH5NO2 Formula Weight: 63.06 Physical Form: Crystalline Density: 1.28 Appearance: White Odour: Ammonia-like Melting Point: ca 116° Beilstein Reference: 3625095 Merck Reference: 14,523 MDL No.: MFCD00013103

Synonyms:  Formic acid ammonium salt
  • CAS Number:540-69-2
  • Formula:HCO₂NH₄
  • MDL Number:MFCD00013103
  • Molecular Weight:63.06 g/mol
  • Merck Index:14,523
  • Melting Pt:∼116 °C
  • Density:1,28
  • Density:1.28
  • Storage Temperature:Omgivande
  • EINECS:208-753-9
Specifications

Specifications

  • Förpackningstyp:
    Plastic bottle for solids
  • Förp.:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.