Ammoniumformat ≥98%
Glass bottle for solids, 50 g
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
Purity: 98% CAS No.: 540-69-2 Molecular Formula: CH5NO2 Formula Weight: 63.06 Physical Form: Crystalline Density: 1.28 Appearance: White Odour: Ammonia-like Melting Point: ca 116° Beilstein Reference: 3625095 Merck Reference: 14,523 MDL No.: MFCD00013103
Synonymer: Formic acid ammonium salt
- CAS-nummer:540-69-2
- Formel:HCO₂NH₄
- MDL-nummer:MFCD00013103
- Molecular Weight:63.06 g/mol
- Merck Index:14,523
- Smält Pt:∼116 °C
- Densitet:1,28
- Densitet:1.28
- Förvaringstemperatur:Omgivande
- EINECS:208-753-9
Specifikationer
Specifikationer
- Förpackningstyp:Glass bottle for solids
- Förp.:50 g



