Order Entry
Sweden
Orders LinkContactUsLinkComponent
Buffert, bruksfärdig

Buffert, bruksfärdig

Technical buffer TEP-10 Trace, 10,01, Plastic bottle, 1 L
Catalog # 662-0270
Supplier:   WTW
Buffert, bruksfärdig
Catalog # 662-0270
Supplier:  WTW
CAS Number:  
Buffert, bruksfärdig
Buffert, bruksfärdig

Technical buffer TEP-10 Trace, 10,01, Plastic bottle, 1 L

CAS Number:  
Price
920,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Every bottle has its own lot reference with the expiration date.

  • Shelf life up to 3 years (unopened)
  • Reference temperature at 25 °C
  • Economic consumption
  • Easiest handling by use of the dosing vessel
  • One litre container with a separate compartment for the sample

Accuracy is ±0,03 pH

Certifications: Traceable to NIST.

Specifications

Specifications

  • Förpackningstyp:
    Plastic bottle
  • Förp.:
    1 L
  • Beskrivning:
    Technical buffer TEP-10 Trace
  • pH-värde:
    10,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.