Urinary system model
Prostate Model, 1/2 natural size
Katalog # BINH1000319
Marketsource
Leverantör: 3B Scientific
Gå till:
Produktdetaljer och dokument
Urinary system all-in-one model on baseboard showing:
- Structures of retroperitoneal cavity
- Large and small pelvis with bones and muscles
- Inferior vena cava
- Aorta with its branches including iliacal vessels
- Upper urinary tract
- Rectum
- Kidney with adrenal gland
The prostate model provides a cross section of the male genital organs showing a healthy prostate with bladder, urethra, testicle, symphysis and rectum.
Specifikationer
- Beskrivning:Prostate Model, 1/2 natural size