Order Entry
Sweden
Orders LinkContactUsLinkComponent
Pulvertratt

Pulvertratt

150 mm, 138 mm, 29 mm
Catalog # 221-0163
Supplier:   VITLAB
Pulvertratt
Catalog # 221-0163
Supplier:  VITLAB
Pulvertratt
Pulvertratt

150 mm, 138 mm, 29 mm

Price
110,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

PP, translucent.

  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable

Used for transferring powders. Also suitable for liquids.

Specifications
  • Tratt, y.d.:
    150 mm
  • Längd:
    138 mm
  • Rör, y.d.:
    29 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.